Learn More
Medchemexpress LLC Selumetinib (sulfate) | 943332-08-9 | 99.6% | C17H17BrClFN4O7S | 1 ML

Supplier: Medchemexpress LLC HY50706A10MM/1ML
Selumetinib (AZD6244) sulfate is a selective, non-ATP-competitive oral MEK1/2 inhibitor. It has an IC50 of 14 nM for MEK1, inhibits ERK1/2 phosphorylation, and can penetrate the blood-brain barrier (BBB).
- Selective, non-ATP-competitive oral MEK1/2 inhibitor
- IC50 of 14 nM for MEK1
- Inhibits ERK1/2 phosphorylation
- Can penetrate the blood-brain barrier (BBB)
- Molecular weight: 555.76
- Appearance: Solid
- Stable under recommended storage conditions
- Identified uses: Laboratory chemicals, manufacture of substances
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.