Learn More
Medchemexpress LLC Selvigaltin (2S) | 00-00-0 | 99.5% | C19H16BrF3N4O4S | 10MG
SDP
Supplier:  Medchemexpress LLC HY147041A10MG
Encompass_Preferred
(2S)-Selvigaltin is the S-enantiomer of selvigaltin, a small-molecule, orally active galectin-3 (Gal-3) inhibitor supplied for research use. The compound is provided with supporting documentation and is characterized by a defined chemical formula, molecular weight, and high purity suitable for in vitro and in vivo studies.
  • S-enantiomer of selvigaltin, offering stereochemically defined activity.
  • Orally active galectin-3 inhibitor used in research applications.
  • High purity (>99%) with documentation including COA and SDS.
  • Characterized chemical formula and molecular weight for reference.
  • Available in small-scale pack sizes suitable for laboratory studies.
Catalog No. 50-002-58482