Learn More
Electron Microscopy Sciences Evactron CombiClean System
SDP
Supplier:  Electron Microscopy Sciences 9100017
Encompass_Preferred
Decontaminate specimens and columns of SEMs and FIBs with the Evactron CombiClean System. The decontaminator combines onboard vacuum cleaning chamber and external PRS (Plasma Radical Source) control in one unified system. The system cleans SEM/TEM samples and SEM chambers from one desktop controller. It stores samples and parts after cleaning. Safar TEM side loaders are used.
Catalog No. 50-192-8223