Learn More
ECM Bioscience Sema-3E and Plexin D1 Receptor Antibody Sampler Kit, ECM Biosciences
Supplier:  ECM Bioscience SK6570
Encompass_Preferred
The Sema-3E and Plexin D1 Receptor kit can be used to monitor the expression of Sema-3E and its receptor Plexin D1. The kit has an N-terminal region Sema-3E antibody, and Plexin D1 antibodies for detecting two regions in the cytoplasmic domain and a region in the extracellular Sema domain. The kit also includes secondary reagent for rabbit polyclonal antibody detection. Applications: WB, E; Species Reactivity: Hu, Rt, Ms
Catalog No. 50-206-8838