Learn More
Medchemexpress LLC Semagacestat (LY450139) | 425386-60-3 | MFCD09970409 | 361.44 g·mol-1 | C19H27N3O4 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1000910MM 1ML
Encompass_Preferred
Semagacestat is a small-molecule γ-secretase inhibitor supplied as a 10 mM solution in DMSO for in vitro research. It is used to study amyloid-β production and γ-secretase-related pathways in neuroscience and Alzheimer's disease research, and is characterized by published molecular and chemical identifiers for traceability.
  • Inhibits γ-secretase activity and reduces amyloid-β peptide production.
  • Provided as a ready-to-use 10 mM solution in DMSO for reproducible dosing.
  • Documented molecular formula and weight for analytical confirmation.
  • Suitable for in vitro and biochemical assays investigating secretase pathways.
  • Available in solid form for custom formulations if required.
Catalog No. 50-002-15309