Learn More
Selleck Chemical LLC Semagacestat (LY450139) 50mg 425386-60-3
SDP
Supplier:  Selleck Chemical LLC S159450MG
Encompass_Preferred
Semagacestat (LY450139) is a ?-secretase blocker for A?42, A?40 and A?38 with IC50 of 10.9 nM, 12.1 nM and 12.0 nM, also inhibits Notch signaling with IC50 of 14.1 nM in H4 human glioma cell. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-561-4