Learn More
Medchemexpress LLC Semaglutide-13C6,15N (TFA) | 96.0% | 4120.53 (free acid) | 500 UG
SDP
Supplier:  Medchemexpress LLC HY114118S3500UG
Encompass_Preferred
Semaglutide-13C6,15N TFA is a 13C- and 15N-labeled form of Semaglutide TFA. This compound is a long-acting, selective, competitive GLP-1R agonist capable of penetrating the blood-brain barrier. It promotes insulin secretion, inhibits gastric emptying and appetite, enhances autophagy, inhibits oxidative stress and apoptosis, and regulates mitochondrial function and lipid metabolism.
  • Used as a tracer.
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Known for lowering blood sugar and reducing weight.
  • Provides neuroprotection, including improving motor function in Parkinson's disease models.
  • Improves hepatic steatosis.
  • Applied in research for type 2 diabetes, obesity, Parkinson's disease, MASLD, and cancer.
Catalog No. 50-004-36843