Learn More
Medchemexpress LLC Semaglutide, FITC labeled | 00-00-0 | 99.8% | 4502.96 | C209H304N46O64S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY114118F5MG
Encompass_Preferred
Semaglutide, FITC labeled is a fluorescein-conjugated semaglutide peptide supplied as a lyophilized powder for use as a fluorescent probe in biochemical and cell-based assays targeting GLP-1 receptor activity. It enables sensitive fluorescence detection and imaging after reconstitution.
Catalog No. 50-002-64036