Learn More
Medchemexpress LLC Semaglutide (TFA) | 99.9% | 4113.58 (free base) | 10 MG
SDP
Supplier:  Medchemexpress LLC HY114118A10MG
Encompass_Preferred
Semaglutide TFA is a long-acting, selective GLP-1R agonist that penetrates the blood-brain barrier. It promotes insulin secretion, inhibits gastric emptying and appetite, and enhances autophagy. It is used in research for conditions like type 2 diabetes, obesity, Parkinson's disease, and fatty liver disease.
  • GLP-1R agonist.
  • Blood-brain barrier penetration.
  • Promotes insulin, inhibits gastric emptying/appetite.
  • Enhances autophagy.
  • Inhibits oxidative stress and apoptosis.
  • Regulates mitochondrial and lipid metabolism.
  • Lowers blood sugar; reduces weight.
  • Neuroprotective effects.
  • Improves hepatic steatosis.
  • Research on neurodegenerative/liver diseases.
Catalog No. 50-004-35835