Learn More
Medchemexpress LLC Semaphorin-3C/SEMA3C 5ug
SDP
Supplier:  Medchemexpress LLC HYP71282A5UG
Encompass_Preferred
Recombinant human Semaphorin-3C (SEMA3C) produced in HEK293 cells with a C-terminal 6xHis tag. The glycosylated protein is supplied as a 0.22 μm-filtered solution formulated for stability in biochemical and cell-based assays and is offered in microgram-scale amounts suitable for receptor-binding, adhesion, and signaling studies. Store frozen and avoid repeated freeze-thaw cycles.
  • Expressed in HEK293 for human-like post-translational modifications.
  • C-terminal 6xHis tag for purification and detection.
  • High purity (>95%) as assessed by reducing SDS-PAGE.
  • Stabilizing formulation containing glycerol, trehalose, arginine, glutamate, and Tween-20.
  • Demonstrated activity in an adhesion assay when immobilized at 10 μg/mL.
  • Recommended storage at -80°C; stable short-term at -20°C.
Catalog No. 50-001-96625