Learn More
Medchemexpress LLC Semaphorin-7A/SEMA7A 100ug | 100ug
SDP
Supplier:  Medchemexpress LLC HYP78035A100UG
Encompass_Preferred
Semaphorin-7A/SEMA7A proteins critically regulate integrin-mediated signaling affecting cell migration and immune responses It promotes focal adhesion complex formation activates PTK2/FAK1 and leads to the phosphorylation of MAPK1 and MAPK3 Semaphorin-7A/SEMA7A Protein Mouse (HEK293 His) is the recombinant mouse-derived Semaphorin-7A/SEMA7A expressed by HEK293 with C-6 His labeled tag
Catalog No. 50-001-87466