Learn More
Medchemexpress LLC Semaphorin-7A/SEMA7A 500ug | 500ug
SDP
Supplier:  Medchemexpress LLC HYP78033500UG
Encompass_Preferred
Semaphorin-7A (SEMA7A) plays a key role in integrin-mediated signaling affecting multiple cellular functions including cell migration and immune responses SEMA7A promotes focal adhesion complex formation activates PTK2/FAK1 and induces phosphorylation of MAPK1 and MAPK3 Semaphorin-7A/SEMA7A Protein Human (HEK293 His) is the recombinant human-derived Semaphorin-7A/SEMA7A protein expressed by HEK293 with C-8 His labeled tag
Catalog No. 50-001-87427