Learn More
Medchemexpress LLC Sematilide hydrochloride | 101526-62-9 | 99.3% | 349.88 g/mol | C14H24ClN3O3S | 1 ML

Supplier: Medchemexpress LLC HY101436A10MM 1ML
Sematilide hydrochloride is a research-grade IKr (delayed rectifier potassium) channel blocker used in electrophysiology and pharmacology studies. The compound is supplied at high purity with molecular data and solubility guidance to support preparation of stock solutions and experimental use.
- High purity suitable for research applications.
- Molecular weight and formula provided for calculation and reporting.
- Solubility and stock-solution guidance available for assay preparation.
- Supplied in forms suitable for aliquoting and storage.
- Intended for in vitro electrophysiology and pharmacology studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.