Learn More
Medchemexpress LLC Sematilide hydrochloride | 101526-62-9 | MFCD11041004 | 99.3% | 349.88 g·mol⁻¹ | C14H24ClN3O3S | 25 MG

Supplier: Medchemexpress LLC HY101436A25MG
Sematilide hydrochloride is the hydrochloride salt of sematilide, a class III antiarrhythmic agent and selective IKr (delayed rectifier K+) channel blocker used in pharmacology research. It inhibits the delayed rectifier K+ current with a reported IC50 of approximately 25 μM and is provided as a high-purity solid for laboratory studies.
- High purity (99.33%).
- Selective IKr (delayed rectifier K+) channel blocker.
- Reported IC50 ≈ 25 μM for delayed rectifier K+ current.
- Molecular weight 349.88 g·mol⁻¹.
- CAS number 101526-62-9.
- Available in milligram-scale pack sizes.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.