Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Semaxanib | 194413-58-6 | MFCD09763655 | 238.29 g·mol⁻1 | C15H14N2O | 250 MG
SDP

Supplier:  Medchemexpress LLC HY10332250MG

Encompass_Preferred

(Z)-Semaxanib, also known as SU5416 or Semaxanib, is a small-molecule tyrosine kinase inhibitor used for laboratory research. It inhibits VEGFR and related receptor tyrosine kinases and has reported cytotoxic activity in hepatic cell lines. The compound is provided as a powder with documented storage recommendations for long-term stability.

  • Small-molecule tyrosine kinase inhibitor targeting VEGFR and c-Met.
  • Shows cytotoxicity in TAMH and HepG2 cells (IC50 6.28 μM and 8.17 μM).
  • Chemical formula C15H14N2O; molecular weight 238.29 g·mol⁻1.
  • Supplied as a powder suitable for research applications.
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-44738


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.