Learn More
Cayman Chemical SemI-synthetIc DrImendIol 1mg
SDP
Supplier:  Cayman Chemical 298201 MG
Encompass_Preferred
A sesquiterpene; inhibits C. albicans, S. aureus, and S. epidermidis biofilm formation (BIC50s = 25.5, 65.1, and 67.1 UG/ml, respectively); inhibits the settlement of C. savignyi and B. improvisus larvae on a petri dish surface (EC50s = 1 and 0.5 UG/ml, respectively)
Catalog No. 50-297-8709