Learn More
Grainger SemiCondctveAbrsnRstntTape 1inx18yd 8mil
Supplier:  Grainger 15D546
Encompass_Preferred
The 3M 15D546 Semi-Conductive Abrasion Resistant Tape is a 1 inch wide by 18 yard long, 8 mil thick tape. It is a rubber splicing tape that is semi-conductive and abrasion resistant. This black tape is rated for 600 volts and can operate between -20F to 194F. It provides insulation and mechanical protection. The tape has quick stick and conforms well to irregular shapes and surfaces. It is designed for splicing and terminating medium and high voltage cable.
Catalog No. 50-274-8728