Learn More
Grainger SemiCondctveAbrsnRstntTape 1inx5yd 8mil
Supplier:  Grainger 15D543
Encompass_Preferred
The Grainger 1" x 5 yard, 8 mil black cloth tape is a general purpose cloth tape ideal for bundling, sealing, splicing, labeling, color coding, and more. It features a pressure sensitive rubber-based adhesive and is made of woven cotton fiber with an acrylic adhesive. This tape is flexible, conformable, and offers a high quick stick. It can adhere to irregular surfaces and has an operating temperature range of 0°F to 200°F. The tape has an unwind adhesion of 28 oz/in, tensile strength of 25 lb/in, and elongation of 8%. It meets UL510 standards for flame retardance. This cloth tape comes in a standard 1" width and 5 yard length roll.
Catalog No. 50-275-1640