Learn More
Grainger SemiCondctveAbrsnRstntTape 2inx5yd 8mil
Supplier:  Grainger 15D544
Encompass_Preferred
The TapeCase Cloth Tape PTFE Slick Surface is a high-quality cloth tape with a PTFE slick surface coating that provides a non-stick and low friction surface. It has a tensile strength of 40 lbs per inch width and adheres to steel, plastic, wood, and more. This 3/4 inch x 60 yard roll of cloth tape is made in the USA and is ideal for use as a non-stick barrier in packaging and processing applications. The PTFE surface resists adhesives and allows for easy, residue-free removal.
Catalog No. 50-275-0293