Learn More
Grainger SemiCondctveAbrsnRstntTape 2inx18yd 8mil
Supplier:  Grainger 15D547
Encompass_Preferred
The TAPECASE Semi-Conductive Abrasion Resistant Tape is a high-quality electrical tape designed for insulating and protecting wires and cables. It features a semi-conductive acrylic adhesive that provides excellent adhesion and abrasion resistance. This 3/4" x 66' tape has a tensile strength of 22 lbs/in width and can withstand temperatures between -18°C to 105°C. It meets UL 510 standard for flame retardance. The tape backing is made of rubber saturated cloth which resists UV rays, oils, acids, alkalis. It provides insulation, moisture sealing, and mechanical protection. This is an ideal tape for insulating electrical splices and repairs.
Catalog No. 50-275-0129