Learn More
Grainger Semiconductor Fuse 350A 170M 700VAC
Supplier:  Grainger 14L635
Encompass_Preferred
The Bussmann Semiconductor Fuse 350 Amp is a high-speed semiconductor fuse designed to protect sensitive electronic equipment. This UL recognized fuse has a voltage rating of 1000VDC and a high interrupting rating of 350,000A. With a super-fast clearing time of less than 8 microseconds, it helps prevent catastrophic failures. The compact cylindrical fuse body has axial leads for easy installation. From one of the most trusted names in circuit protection, this fuse provides reliable overload and short circuit protection for semiconductors and other sensitive components.
Catalog No. 50-275-1268