Learn More
Grainger Semiconductor Fuse 630A 170M Indicating
Supplier:  Grainger 14L637
Encompass_Preferred
The BUSSMANN Semiconductor Fuse 630-A Amps is a fast-acting, high interrupting rating semiconductor fuse designed to protect sensitive electronic equipment. This UL recognized fuse has a voltage rating of 250VAC and features a ceramic body construction with nickel-plated brass endcaps and blade terminals. The fuse measures 1-1/4" long x 1/4" wide and has a maximum opening of 630 amps at 250 volts. With high surge withstand capability and low let-through values, this semiconductor fuse provides reliable overload and short circuit protection for semiconductors and other sensitive electronic components. The compact design allows for high density board mounting.
Catalog No. 50-275-1269