Learn More
Perkin Elmer US LLC Semi-Demountable Spacers - Assorted
Supplier:  Perkin Elmer US LLC L1270421
Encompass_Preferred
This packet includes the following rectangular teflon spacers: L1270402, 0.05 mm thick L1270404, 0.1 mm thick L1270408, 0.2 mm thick L1270409, 0.5 mm thick L1270410, 1 mm thick
Catalog No. 50-211-1909