Learn More
Cayman Chemical SemiQuantitative Biomarker EL
SDP
Supplier:  Cayman Chemical 100086595 EA
Encompass_Preferred
This assay contains a set of reagents to be used together with a suitable Biosense primary antibody for semi-quantitative detection of biomarkers such as Vtg, CYP1A, Zrp, and MT in samples from fish.
Catalog No. 50-288-6532