Learn More
Medchemexpress LLC Semisynthetic protoberberine derivative | 908001-51-4 | 98.0% | 562.49 | C31H32BrNO4 | 10 MG

Supplier: Medchemexpress LLC HY16345910MG
HWY-289 is a semisynthetic protoberberine derivative with broad-spectrum antifungal activity against phytopathogenic fungi, particularly Botrytis cinerea. It alters fungal mycelium morphology and cellular structure, and impairs energy metabolism by reducing ATP content, ATPase activities, and key TCA cycle enzymes. Supplied as a purified research reagent for laboratory use.
- Broad-spectrum antifungal activity, active vs Botrytis cinerea (EC50 = 1.34 μg/mL).
- Alters mycelial morphology and internal cell structure.
- Reduces ATP content, ATPase activity, and TCA cycle enzyme activities.
- Chemical identifiers: CAS 908001-51-4; formula C31H32BrNO4; molecular weight 562.49.
- Purity 98.0%.
- Available in multiple sizes including 10 MG.
- Storage: 4°C sealed, away from moisture and light; in solvent: -80°C (6 months), -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.