Learn More
Medchemexpress LLC HY-137450 5mg Medchemexpress, Senaparib CAS:1401682-78-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1374505MG
Encompass_Preferred
Medchemexpress, HY-137450 5mg Senaparib CAS:1401682-78-7 Senaparib (IMP4297) is a highly potent, selective and orally active PARP1/2 inhibitor. Senaparib (IMP4297) exhibits strong antitumor activity in animal models. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1489