Learn More
ATCC Seneca Valley virus (ATCC VR-1989)
Supplier:  ATCC VR1989
Encompass
Seneca Valley virus is a non-enveloped, positive-sense, single-stranded RNA picornavirus (Senecavirus A). The ATCC VR-1989 strain is an authenticated isolate provided for laboratory study of viral biology, pathogenesis, oncolytic activity, and veterinary surveillance.
  • Non-enveloped, positive-sense, single-stranded RNA virus.
  • Belongs to the family Picornaviridae, genus Senecavirus.
  • Used in studies of viral pathogenesis and oncolytic properties.
  • Relevant to veterinary surveillance as a porcine pathogen.
  • Provided as authenticated culture material for laboratory use.
Catalog No. NC3360038