Learn More
Sigma Aldrich Fine Chemicals Biosciences Senescence Cells Histochem
Supplier:  Sigma Aldrich Fine Chemicals Biosciences CS00301KT
Encompass
The Senescence Cells Histochemical Staining Kit contains all the reagents required for identifying senescent cells using an assay based on a histochemical stain for b-galactosidase activity at pH 6. b-galactosidase activity is detectable in senescent cells but not in quiescent immortal or tumor cells.
Catalog No. 50-290-5794