Learn More
Selleck Chemical LLC Senexin A 25mg 1366002-50-7
SDP
Supplier:  Selleck Chemical LLC S852025MG
Encompass_Preferred
Senexin A is a potent and selective inhibitor of CDK8 and its nearest relative, CDK19 with Kd values of 0.83 ?M and 0.31 ?M for CDK8 and CDK19 ATP site binding, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1698628