Learn More
Medchemexpress LLC Senicapoc | 289656-45-7 | 99.9% | C20H15F2NO | 50 MG
SDP
Supplier:  Medchemexpress LLC HY5069450MG
Encompass_Preferred
Senicapoc (ICA-17043) is a potent and selective Gardos channel (Ca2+-activated K+ channel; KCa3.1) blocker. It blocks Ca2+-induced rubidium flux from human RBCs and inhibits RBC dehydration. It has been shown to decrease Gardos channel activity and increase red cell K+ content in vivo. It also reduces airway hyperresponsiveness and vascular remodelling in certain models.
  • Potent and selective Gardos channel (Ca2+-activated K+ channel; KCa3.1) blocker
  • Blocks Ca2+-induced rubidium flux from human red blood cells
  • Inhibits red blood cell dehydration
  • Decreases Gardos channel activity and increases red cell K+ content
  • Reduces airway hyperresponsiveness and vascular remodelling
Catalog No. 50-003-58761