Learn More
Medchemexpress LLC Senicapoc | 5 MG
SDP
Supplier:  Medchemexpress LLC HY506945MG
Encompass_Preferred
Senicapoc (ICA-17043) is a potent and selective Gardos channel (Ca2+-activated K+ channel; KCa3.1) blocker. It has an IC50 of 11 nM for this blockade and also inhibits Ca2+-induced rubidium flux from human RBCs with an IC50 value of 11 nM. Furthermore, it inhibits RBC dehydration with an IC50 of 30 nM.
  • Potent and selective Gardos channel (Ca2+-activated K+ channel; KCa3.1) blocker
  • Blocks Ca2+-induced rubidium flux from human RBCs
  • Inhibits RBC dehydration
Catalog No. 50-003-59128