Learn More
Selleck Chemical LLC Senicapoc-E0501-5MG
SDP
Supplier:  Selleck Chemical LLC E05015MG
Encompass_Preferred
Senicapoc is a Gardos channel(Ca2 -activated K channel KCa3 1) blocker with an IC50 of 11 nM and produces dose-dependent increases in hemoglobin and decreases in markers of hemolysis Senicapoc blocks Ca2 -induced rubidium flux from human RBCs with an IC50 value of 11 nM and inhibits RBC dehydration with IC50 of 30 nM
Catalog No. 50-313-7260