Learn More
Medchemexpress LLC HY-P0187 1mg Medchemexpress, Senktide CAS:106128-89-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP01871MG
Encompass_Preferred
Medchemexpress, HY-P0187 1mg Senktide CAS:106128-89-6 Senktide is a tachykinin NK 3 receptor agonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4045