Learn More
Medchemexpress LLC Sennoside C | 37271-16-2 | 848.76 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN19721MG
Encompass_Preferred
Sennoside C is an orally active component of the traditional Chinese medicine Sennae Folium. It can bind to human lysozyme and inhibit its amyloid fibrosis (IC50: 186.20 μM). It also possesses purgative activity and is used in the research of diarrhea and anti-amyloid related diseases.
  • Solid appearance
  • Color is white to yellow
  • Structure classification: Quinones, anthraquinones, phenols, polyphenols
  • Initial source: Plants, leguminosae, cassia angustifolia
Catalog No. 50-004-15686