Learn More
Medchemexpress LLC Sennoside D | 37271-17-3 | 848.76 | 1 MG

Supplier: Medchemexpress LLC HYN19731MG
Sennoside D is an anthraquinone glycoside, found in leaves and pods of Senna (Cassia angustifolia). It is a solid, light yellow to brown in appearance, with a purity of 98.36% and a molecular formula of C42H40O19.
- Store at 4°C, sealed, away from moisture and light.
- When in solvent, store at -80°C for 6 months or -20°C for 1 month, sealed, away from moisture and light.
- Purity: 98.36%.
- Appearance: Solid.
- Color: Light yellow to brown.
- Molecular formula: C42H40O19.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.