Learn More
Medchemexpress LLC SENP2-IN-1 | 2986220-46-4 | C32H29N3O5S2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1513585MG
Encompass_Preferred
SENP2-IN-1 (compound 77) is a selective SENP2 inhibitor. It exhibits inhibitory activities with IC₅₀ values of 1.3 μM for SENP1, 0.69 μM for SENP2, and 22.7 μM for SENP5. This compound is suitable for research related to cancer.
  • Selective SENP2 inhibitor
  • Exhibits inhibitory activities with IC₅₀ values of 1.3 μM for SENP1, 0.69 μM for SENP2, and 22.7 μM for SENP5
  • Suitable for cancer research
  • Applicable to digestive system disease, liver cancer, and digestive system cancer research
  • Used in cancer targeted therapy, cancer metabolism, and metastasis research
Catalog No. 50-003-75807