Learn More
Bachem Z-Arg-AMC
SDP
Supplier:  Bachem 4001519.1000
Encompass_Preferred
Z-Arg-AMC Hydrochloride, 62037-44-9, C24H27N5O5, Mr 465.51, 1g. Sensitive fluorogenic substrate for trypsin, soybean trypsin-like enzyme, and papain.
Catalog No. 50-194-7065