Learn More
AnaSpec SensoLyte Blue Glucocerebrosidase Activity Assay Kit, Fluorimetric Detection, 100 Assays (96-Well Plate)
Supplier:  AnaSpec AS72258
Encompass
The SensoLyte Blue Glucocerebrosidase Activity Assay Kit is a fluorometric kit optimized for measuring GBA enzyme activity, crucial for diagnosing Gaucher disease and related disorders. It provides a reliable, high-speed assay with a total reaction time of one hour, utilizing a substrate that yields bright blue fluorescence. Ideal for research applications, it comes with sufficient reagents for 100 assays in a 96-well format, ensuring ease of use for compound screening and other research needs.
Catalog No. NC3037674