Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Corporation Sentry Guard Holder, Universal, 4.6 mm ID, 20 mm Length

Supplier:  Waters Corp WAT046910

Encompass_Preferred

This Sentry Guard Holder is designed for use with cartridge and guard columns. It features a universal design for broad compatibility.

  • Holder kit for cartridge and guard columns.
  • Universal design.
  • Compatible with 4.6 mm inner diameter columns.
  • Designed for 20 mm length columns.
  • Includes a U-connector.

Catalog No. 50-818-722


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.