Learn More
Grainger Sentry Plus Shirt Black Neck 17 in
Supplier:  Grainger 14F636
Encompass_Preferred
The Grainger 14F636 is a Westward brand 1/4-inch drive socket set that includes both SAE and metric sockets. The set contains 15 sockets ranging in size from 4mm to 14mm and 1/8 inch to 9/16 inch. The chrome vanadium sockets have a matte finish and feature a knurled design for easy gripping. The set comes packaged in a molded case for convenient storage and portability. The Westward tools are designed for automotive and machinery applications. This socket set provides both SAE and metric sockets in a portable case for mobile mechanics.
Catalog No. 50-275-2159