Learn More
Medchemexpress LLC Seocalcitol | 134404-52-7 | 99.3% | 10 MG

Supplier: Medchemexpress LLC HY3234110MG
Seocalcitol is a synthetic vitamin D analog provided as a white to off-white solid for research use. It binds the vitamin D receptor with high affinity and is used to study VDR-mediated signaling, antiproliferative effects, and calcium-related pathways in cellular and tissue assays. For research use only; not for human or veterinary use.
- High purity (99.3%).
- White to off-white solid form.
- Supplied as 10 MG quantity.
- Molecular formula C30H46O3; molecular weight 454.68 g/mol.
- Binds vitamin D receptor with Kd of 0.27 nM (MG-63 cells).
- Protect from light and store under nitrogen; recommend 4°C storage; in solvent: -80°C for up to 6 months or -20°C for up to 1 month.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.