Learn More
Waters Corp Sep-Pak C8 Vac RC Cartridge, 100 mg Sorbent, 37 to 55 μm
Supplier: Waters Corp WAT043415
This silica-based bonded phase cartridge offers moderate hydrophobicity, suitable for methods requiring less retention than C18. It is typically used for applications such as the analysis of drugs and their metabolites in serum, plasma, or urine, and peptides in serum or plasma.
- Features a C8 chemistry.
- Endcapped for improved peak shape and reproducibility.
- Designed as a Vac RC Cartridge.
- Compatible with mass spectrometry.
- Operates in reversed phase mode.
- Offers a particle size range of 37 to 55 μm.
- Has a pore size of 125 Å.
- Utilizes a silica sorbent substrate.
- Contains 100 mg of sorbent.
- Not water wettable.
- Supports a pH range of 2 to 8.
- Has a hold up volume of 0.25 mL.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.