Learn More
Cole-Parmer DWK Life Sciences (Kimble) 29048F500 Separatory Funnel, PTFE Stopcock, 500 mL 4/cs
Supplier:  Cole-Parmer 3450548
Encompass_Preferred
Funnel is manufactured with stem ID large enough that allows separation when stopcock is closed; allowing a more complete separation and eliminates the need for emptying the funnel.
Catalog No. NC2160032