Learn More
Sepax Technologies Inc Sepax HPLC Column Antibodix WCX, NP10, 21.2 x 50 mm
SDP
Supplier:  Sepax Technologies Inc 602NP1021205
Encompass
Antibodix columns are specially designed for high resolution, high efficiency and high recovery separations of antibodies. The packing support is composed of a rigid, spherical, highly cross-linked poly(styrene divinylbenzene) (PS/DVB) non-porous bead. The non-porous resin has particle size of 1.7, 3, 5 and 10 µm. The PS/DVB resin surface is grafted with a highly hydrophilic, neutral polymer thin layer with the thickness in the range of nanometer. On the top of the hydrophilic layer, weak cation-exchange functional groups are attached via a proprietary chemistry, resulting in high capacity ion-exchange layer.
Catalog No. 50-233-8048