Learn More
Sepax Technologies Inc Sepax HPLC Column Bio-C4, 10 µm, 300 Å, 10 x 150 mm
SDP
Supplier:  Sepax Technologies Inc 11004910015
Encompass
Monomeric and fully endcapped Bio-C4 packing is bonded with butyl groups that lead to moderate hydrophobicity. Bio-C4 packings of 300 Å pore size selection are ideal for high resolution mapping of peptides and separation of intact proteins, and natural and synthetic peptides.
Catalog No. 50-233-8351