Learn More
Sepax Technologies Inc Sepax HPLC Column GP-C4, 5 µm, 120 Å, 2.1 x 250 mm
SDP
Supplier:  Sepax Technologies Inc 1090452125
Encompass
Monomeric and fully endcapped GP-C4 packing is bonded with butyl groups that lead to moderate hydrophobicity. GP-C4 columns have unique selectivity and peak symmetry with moderate retention for separations of acidic, neutral and basic organic compounds, such as pharmaceuticals, peptides, and organic acids.
Catalog No. 50-233-8491