Learn More
Sepax Technologies Inc Sepax HPLC Column GP-Phenyl, 5 µm, 120 Å, 2.1 x 100 mm
SDP
Supplier:  Sepax Technologies Inc 1113652110
Encompass
GP-Phenyl packing materials are bonded with propyl phenyl groups that enable special interaction with ring structured compounds. The monomeric bonding chemistry gives high efficiency and high resolution separations. GP-Phenyl phase is suitable for separations of acidic, neutral and basic organic compounds, as well as pharmaceuticals.
Catalog No. 50-233-6629