Learn More
Sepax Technologies Inc Sepax HPLC Column HP-Amino, 5 µm, 120 Å, 4.6 x 100 mm
SDP
Supplier:  Sepax Technologies Inc 1153054610
Encompass
Synthesized with polymeric chemistry, HP-Amino phase is bonded with aminopropyl functional group. HP-Amino phase is compatible with versatile mobile phases from non-aqueous solvents, such as hexane/ethyl acetate and chloroform/methanol, to aqueous solutions. It is recommended for separations of sugars, nucleotides, basic organic compounds, as well as the pharmaceuticals.
Catalog No. 50-233-6917