Learn More
Sepax Technologies Inc Sepax HPLC Column Polar-Imidazole, 5 µm, 120 Å, 2.1 x 250 mm
SDP
Supplier:  Sepax Technologies Inc 1353352125
Encompass
Utilizing highest purity and enhanced mechanical stability silica and pure bonding reagents, Polar-Imidazole phase is bonded with imidazole functional groups. With a polar and N-containing ring structure, Polar-Pyridine phase is a weak base and adequate for many applications, such as pharmaceuticals, peptides, sugars, and nucleotides.
Catalog No. 50-233-6779