Learn More
Sepax Technologies Inc Sepax HPLC Column SRT-10 SEC-100, 10 µm, 21.2 x 300 mm
SDP
Supplier:  Sepax Technologies Inc 22510021230
Encompass
Utilizing proprietary surface technologies, SRT-10 and SRT-10C SEC phases are made of uniform, hydrophilic, and neutral nanometer thick films chemically bonded on high purity and mechanically stabilized 10 µm silica. The well-controlled surface chemistry results in excellent lot-to-lot reproducibility. Our unique bonding chemistry, coupled with the maximized bonding density, allows SRT-10 and SRT-10C SEC phases to provide high stability and negligible non-specific interactions. SRT-10 and SRT-10C packings have large pore volume, resulting in high separation resolution. Typical applications for SRT-10 and SRT-10C columns include separation and detection of biological molecules and water-soluble polymers in aqueous buffers.
Catalog No. 50-233-6561
Only null left
Add to Cart
Request a Quote